missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAN2A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69054
This item is not returnable.
View return policy
Description
MAN2A2 Polyclonal antibody specifically detects MAN2A2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| MAN2A2 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| alpha-mannosidase 2x, alpha-mannosidase IIx, MAN IIx, MANA2X, mannosidase, alpha type II-X, mannosidase, alpha, class 2A, member 2, mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase, manosidase, alpha-, type II, isozyme X | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TLPSSVRIYLHGRQLSVSRHEAFPLRVIDSGTSDFALSNRYMQVWFSGLTGLLKSIRRVDEEHEQQVDMQVLVYGTRTSKD | |
| 100 μg | |
| Cellular Markers, Golgi Apparatus Markers | |
| 4122 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction