missing translation for 'onlineSavingsMsg'
Learn More

Mandelic acid hydrazide, 97%

Product Code. 11312557
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312557 25 g 25g
11302557 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312557 Supplier Thermo Scientific Alfa Aesar Supplier No. L11653.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2443-66-5 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2443-66-5
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00038133
InChI Key FWTGUGVETHVGTL-UHFFFAOYSA-N
Synonym mandelic acid hydrazide, mandelhydrazide, mandelhydrazine, mandelic acid, hydrazide, mandelic hydrazide, hydroxy-phenyl-acetic acid hydrazide, 2-???-1-phenylethan-1-ol, mandelohydrazide, asischem d13400, akos bc-1667
PubChem CID 73126
IUPAC Name 2-hydroxy-2-phenylacetohydrazide
SMILES C1=CC=C(C=C1)C(C(=O)NN)O

Specifications

Melting Point 128°C to 131°C
Quantity 25 g
Beilstein 2096197
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material Mandelic acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.