missing translation for 'onlineSavingsMsg'
Learn More

Mangiferin, MedChemExpress

Product Code. 16478938
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478938 10 mg 10mg
16474807 50 mg 50mg
16439046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478938 Supplier Medchem Express Supplier No. HYN0290
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mangiferin is a Nrf2 activator. Mangiferin suppresses nuclear translocation of the NF-κB subunits p65 and p50. Mangiferin exhibits antioxidant, antidiabetic, antihyperuricemic, antiviral, anticancer and antiinflammatory activities.

Mangiferin is a Nrf2 activator. Mangiferin suppresses nuclear translocation of the NF-κB subunits p65 and p50. Mangiferin exhibits antioxidant, antidiabetic, antihyperuricemic, antiviral, anticancer and antiinflammatory activities.

Specifications

Chemical Name or Material Mangiferin
Physical Form Solid
CAS 4773-96-0
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H18O11
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 31.25 mg/mL (73.99 mM; ultrasonic and warming and heat to 80°C)
SMILES O[C@@H]([C@H](O)[C@@H](CO)O1)[C@@H](O)[C@@H]1C2=C(O)C3=C(C=C2O)OC4=CC(O)=C(O)C=C4C3=O
Molecular Weight (g/mol) 422.34
Formula Weight 422.34
Percent Purity 98.34%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.