missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannose Phosphate Isomerase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 539.00 €
Specifications
| Antigen | Mannose Phosphate Isomerase |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18495711
|
Novus Biologicals
NBP1-85787-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286957
|
Novus Biologicals
NBP1-85787 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mannose Phosphate Isomerase Polyclonal specifically detects Mannose Phosphate Isomerase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Mannose Phosphate Isomerase | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4351 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GSNSEVARLLASSDPLAQIAEDKPYAELWMGTHPRGDAKILDNRISQKTLSQWIAENQDSLGSKVKDTFNGNLPFLFKVLSVETPLSIQAHPNKELAEKLHLQAPQHYPDANHKPEMAIALTPFQGLCG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| CDG1B, EC 5.3.1, EC 5.3.1.8, FLJ39201, mannose phosphate isomerase, mannose-6- phosphate isomerase, mannose-6-phosphate isomerase, Phosphohexomutase, Phosphomannose isomerase, phosphomannose isomerase 1, PMI, PMI1phosphohexomutase | |
| MPI | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title