missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ MAP 2a,b (Microtubule-Associated Protein) Ab-2, Mouse Monoclonal Antibody

Product Code. 12660697 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Unit Size:
0.1mL
1mL
500µL
7mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12660697 100 μL Concentrated 0.1mL
12670697 500 μL Concentrated 500µL
12650697 1 mL Concentrated 1mL
12640697 7 mL Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12660697 Supplier Epredia™ Supplier No. MS249S0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ MAP 2a,b (Microtubule-Associated Protein) Ab-2, Mouse Monoclonal Antibody.

Microtubule Associated Protein 2 exists in two high molecular weight forms (2a & 2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. Ab-2 is highly specific to MAP2a,b and shows no cross-reaction with MAP1, MAP5, tubulin, or tau. Ab-2 reacts with dendrites and cell bodies of neurons but not with the neuronal processes. It does not react with the low molecular weight (70kDa) form (2c) of MAP2. It is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's.

Host Species: Mouse

Clone: AP20

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 997-1332

Immunogen: Bovine brain microtubule associated protein 2

Molecular Weight: ∽280kDa Doublet

Positive Control: Human glioblastoma T98G cells or brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP 2a,b (Microtubule-Associated Protein) Ab-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone AP20
Conjugate Unconjugated
Host Species Mouse
Immunogen Bovine brain microtubule associated protein 2
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.