missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ MAP 2a,b (Microtubule-Associated Protein) Ab-2, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Microtubule Associated Protein 2 exists in two high molecular weight forms (2a & 2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. Ab-2 is highly specific to MAP2a,b and shows no cross-reaction with MAP1, MAP5, tubulin, or tau. Ab-2 reacts with dendrites and cell bodies of neurons but not with the neuronal processes. It does not react with the low molecular weight (70kDa) form (2c) of MAP2. It is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's.
Host Species: Mouse
Clone: AP20
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: aa 997-1332
Immunogen: Bovine brain microtubule associated protein 2
Molecular Weight: ∽280kDa Doublet
Positive Control: Human glioblastoma T98G cells or brain
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MAP 2a,b (Microtubule-Associated Protein) Ab-2 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | AP20 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Bovine brain microtubule associated protein 2 |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Research Discipline | Neuroscience |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?