Learn More
MAP2 Antibody (CL5420) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61417AF647
Product Code. 30290188
  • 748.00 € / 0.10mL
Description
MAP2 Monoclonal antibody specifically detects MAP2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Single Cell Western,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MAP2
Monoclonal
Alexa Fluor 647
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
This antibody was developed using a recombinant protein derived from P11137, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
4133
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Single Cell Western, Immunocytochemistry/Immunofluorescence
CL5420
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified