missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K12 binding inhibitory protein 1 Antibody (8G3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42994
This item is not returnable.
View return policy
Description
MAP3K12 binding inhibitory protein 1 Monoclonal antibody specifically detects MAP3K12 binding inhibitory protein 1 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| MAP3K12 binding inhibitory protein 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 8G3 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunoprecipitation 1:200 - 1:1000 | |
| MAP3K12 binding inhibitory protein 1, MAP3K12-binding inhibitory protein 1, MAPK upstream kinase-binding inhibitory protein, MUK-binding inhibitory protein | |
| MAP3K12 binding inhibitory protein 1 - (Uniprot# Q9NS73) | |
| 100 μg | |
| Protein Kinase | |
| 51562 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction