missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MAP3K8/Tpl2/COT Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
250µg
Specifications
Specifications
| Gene ID (Entrez) | 1326 |
| Species | Human |
| Purity | >85%, by HPLC |
| Concentration | 2.5 mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-62178 |
| Gene Alias | Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2 |
| Gene Symbol | MAP3K8 |
| Quantity | 0.25 mg |
| Research Category | Angiogenesis, Cancer, Protein Kinase |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction