missing translation for 'onlineSavingsMsg'
Learn More

Maprotiline hydrochloride, 99%

Product Code. 15414729
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414729 1 g 1g
15424729 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414729 Supplier Thermo Scientific Alfa Aesar Supplier No. J62321.03
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective norepinephrine uptake inhibitor | CAS: 10347-81-6 | C20H24ClN | 313.87 g/mol

Maprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.

Solubility
Soluble to 50mg/ml in water. Soluble in ethanol at 10mM

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10347-81-6
Molecular Formula C20H24ClN
Molecular Weight (g/mol) 313.87
MDL Number MFCD00079464
InChI Key NZDMFGKECODQRY-UHFFFAOYSA-N
Synonym maprotiline hydrochloride, maprotiline hcl, ludiomil, psymion, maprotilline hcl, ciba 34276 ba, unii-7c8j54pvfi, deprilept, 9-gamma-methylaminopropyl-9,10-dihydro-9,10-ethanoanthracene hydrochloride
PubChem CID 71478
IUPAC Name hydrogen methyl(3-{tetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-1-yl}propyl)amine chloride
SMILES [H+].[Cl-].CNCCCC12CCC(C3=CC=CC=C13)C1=CC=CC=C21

Specifications

Melting Point 230°C to 232°C
Color White
Quantity 1 g
Beilstein 5463242
Merck Index 14,5748
Solubility Information Soluble to 50mg/ml in water; Soluble in ethanol at 10mM
Formula Weight 313.87
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Maprotiline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.