missing translation for 'onlineSavingsMsg'
Learn More

Maprotiline hydrochloride, MedChemExpress

Product Code. 16492136
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492136 500 mg 500mg
16428316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492136 Supplier Medchem Express Supplier No. HYB0444
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Maprotiline hydrochloride is a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant.

Maprotiline hydrochloride is a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant.

Specifications

Chemical Name or Material Maprotiline hydrochloride
CAS 10347-81-6
Color White
Purity Grade Notes Research
Molecular Formula C20H24ClN
Quantity 500 mg
Solubility Information DMSO : 83.33 mg/mL (265.50 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2.2 mg/mL (7.01 mM; Need ultrasonic)
SMILES CNCCCC1(CC2)C3=CC=CC=C3C2C4=CC=CC=C14.Cl
Molecular Weight (g/mol) 313.86
Formula Weight 313.86
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.