missing translation for 'onlineSavingsMsg'
Learn More

Mardepodect hydrochloride, MedChemExpress

Product Code. 16473977
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473977

Brand: Medchem Express HY50098A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mardepodect hydrochloride (PF-2545920 hydrochloride) is a potent, orally active and selective PDE10A inhibitor with an IC50 of 0.37 nM, with >1000-fold selectivity over other PDEs. Mardepodect hydrochloride can cross the blood-brain barrier.

Mardepodect hydrochloride (PF-2545920 hydrochloride) is a potent, orally active and selective PDE10A inhibitor with an IC50 of 0.37 nM, with >1000-fold selectivity over other PDEs. Mardepodect hydrochloride can cross the blood-brain barrier.

Specifications

Chemical Name or Material Mardepodect hydrochloride
CAS 2070014-78-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H21ClN4O
Quantity 50 mg
Synonym PF-2545920 hydrochloride
Solubility Information DMSO : 25 mg/mL (58.29 mM; Need ultrasonic)
SMILES CN1N=C(C2=CC=C(OCC3=NC4=CC=CC=C4C=C3)C=C2)C(C5=CC=NC=C5)=C1.Cl[H]
Molecular Weight (g/mol) 428.91
Formula Weight 428.91
Percent Purity 95.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.