missing translation for 'onlineSavingsMsg'
Learn More

Maropitant Citrate Salt, TRC

Product Code. 17901275
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901275 1 mg 1mg
17922125 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17901275 Supplier Toronto Research Chemicals Supplier No. TRCM1973001MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862543-54-2
Molecular Formula C32 H40 N2 O . C6 H8 O7
Molecular Weight (g/mol) 660.8
Synonym (2S-cis)-N-[[5-(1,1-Dimethylethyl)-2-methoxyphenyl]methyl]-2-(diphenylmethyl)-1-azabicyclo[2.2.2]octan-3-amine Citrate Salt, (2S,3S)-2-Benzhydryl-N-(5-tert-butyl-2-methoxybenzyl)quinuclidin-3-amine Citrate Salt, Cerenia
SMILES CC(C)(C)C1=CC(=C(C=C1)OC)CN[C@@H]2[C@@H](N3CCC2CC3)C(C4=CC=CC=C4)C5=CC=CC=C5.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C32H40N2O.C6H8O7/c1-32(2,3)27-15-16-28(35-4)26(21-27)22-33-30-25-17-19-34(20-18-25)31(30)29(23-11-7-5-8-12-23)24-13-9-6-10-14-24;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-16,21,25,29-31,33H,17-20,22H2,1-4H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/t30-,31-;/m0./s1
Percent Purity >95%
Chemical Name or Material Maropitant Citrate Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.