missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MART-1 / MLANA Rabbit, Clone: MLANA/1409R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 2315-RBM9-P0
This item is not returnable.
View return policy
Specifications
| MART-1 / MLANA | |
| Cytoplasmic | |
| MLANA/1409R | |
| Unconjugated | |
| NP_005502 | |
| Rabbit | |
| 20-22kDa (doublet) | |
| Protein A/G Purified | |
| RUO | |
| 2315 | |
| IgG κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| MLANA | |
| Recombinant human full-length MLANA protein | |
| SK-MEL-13 and SK-MEL-19 Melanoma cell lines; Melanomas | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction