missing translation for 'onlineSavingsMsg'
Learn More

Maryland Pesticide Mixture 2 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30324244
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324244 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324244 Supplier Dr. Ehrenstorfer Supplier No. DRES50000141AL
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bifenazate∣Bifenthrin∣Boscalid∣Chlorpyrifos∣Cyfluthrin∣Diazinon∣Fipronil∣Flurprimidol∣Hexythiazox∣Metalaxyl∣Paclobutrazol∣Permethrin∣Phosmet∣Piperonyl butoxide∣Pyrethrins∣Trifloxystrobin
CAS 149877-41-8, 82657-04-3, 188425-85-6, 2921-88-2, 68359-37-5, 333-41-5
Analyte or Component Names Bifenazate∣Bifenthrin∣Boscalid∣Chlorpyrifos∣Cyfluthrin∣Diazinon∣Fipronil∣Flurprimidol∣Hexythiazox∣Metalaxyl∣Paclobutrazol∣Permethrin∣Phosmet∣Piperonyl butoxide∣Pyrethrins∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Bifenazate 1000 μg/mL∣Bifenthrin 1000 μg/mL∣Boscalid 1000 μg/mL∣Chlorpyrifos 1000 μg/mL∣Cyfluthrin 1000 μg/mL∣Diazinon 1000 μg/mL∣Fipronil 1000 μg/mL∣Flurprimidol 1000 μg/mL∣Hexythiazox 1000 μg/mL∣Metalaxyl 1000 μg/mL∣Paclobutrazol 1000 μg/mL∣Permethrin 1000 μg/mL∣Phosmet 1000 μg/mL∣Piperonyl butoxide 1000 μg/mL∣Pyrethrins 1000 μg/mL∣Trifloxystrobin 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.