missing translation for 'onlineSavingsMsg'
Learn More

Masitinib, MedChemExpress

Product Code. 16493708
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493708 10 mg 10mg
16460097 50 mg 50mg
16434786 100 mg 100mg
16490946 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493708 Supplier Medchem Express Supplier No. HY10209
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Masitinib (AB1010) is a potent, orally bioavailable, and selective inhibitor of c-Kit (IC50=200 nM for human recombinant c-Kit). It also inhibits PDGFRα/β (IC50s=540/800 nM), Lyn (IC50= 510 nM for LynB), Lck, and, to a lesser extent, FGFR3 and FAK. Masitinib (AB1010) has anti-proliferative, pro-apoptotic activity and low toxicity.

Masitinib (AB1010) is a potent, orally bioavailable, and selective inhibitor of c-Kit (IC50=200 nM for human recombinant c-Kit). It also inhibits PDGFRα/β (IC50s=540/800 nM), Lyn (IC50= 510 nM for LynB), Lck, and, to a lesser extent, FGFR3 and FAK. Masitinib (AB1010) has anti-proliferative, pro-apoptotic activity and low toxicity.

Specifications

Chemical Name or Material Masitinib
Physical Form Solid
CAS 790299-79-5
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C28H30N6OS
For Use With (Application) Cancer-Kinase/protease
Synonym AB1010
Solubility Information DMSO : 100 mg/mL (200.55 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=CC=C(C(NC2=NC(C3=CC=CN=C3)=CS2)=C1)C)C4=CC=C(CN5CCN(CC5)C)C=C4
Molecular Weight (g/mol) 498.64
Formula Weight 498.64
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.