missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mast Cell Chymase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18189-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene product is a chymotryptic serine proteinase that belongs to the peptidase family S1. It is expressed in mast cells and thought to function in the degradation of the extracellular matrix, the regulation of submucosal gland secretion, and the generation of vasoactive peptides. In the heart and blood vessels, this protein, rather than angiotensin converting enzyme, is largely responsible for converting angiotensin I to the vasoactive peptide angiotensin II. Angiotensin II has been implicated in blood pressure control and in the pathogenesis of hypertension, cardiac hypertrophy, and heart failure. Thus, this gene product is a target for cardiovascular disease therapies. This gene maps to 14q11.2 in a cluster of genes encoding other proteases.Specifications
| Mast Cell Chymase | |
| Polyclonal | |
| Unconjugated | |
| CMA1 | |
| Alpha chymase, chymase, chymase 1, mast cell, CMA1, CYH, CYM, Mast Cell Chymase, Mast cell protease I, Mcpt 5, Mcpt5, MCT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1215, 17228, 25627 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21844, P23946, P50339 | |
| Cma1 | |
| Mast Cell Chymase Fusion Protein Ag12909 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction