Learn More
Mast Cell Marker Antibody (MCG35) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50484-0.025ml
 View more versions of this product
Product Code. 18674666
  • 201.00 € / 0.03mL
Description
Mast Cell Marker Monoclonal antibody specifically detects Mast Cell Marker in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mast Cell Marker
Monoclonal
1 mg/mL
Immunohistochemistry 1:10 - 1:500
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Immunohistochemistry
MCG35
Unconjugated
PBS
Spleen cells and bone marrow cells (erythrocyte depleted) from a patient with systemic mastocytosis. The bone marrow preparation consisted of 50% typical mast cells.
0.025 mL
Cardiovascular Biology
Human
Purified