missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- ideal for storage of active agents, patient samples, biomolecules and reagents and cultivation of bacteria or yeasts
- Uniform external dimensions and tolerances
- Alphnumeric well coding
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
Specifications
Specifications
| Certifications/Compliance | DIN ISO 9001 |
| Certified | RNase-/DNase, pyrogen free |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Packaging | 1/Pk., 50/Cs. |
| Color | Clear |
| Product Type | 96-well Microplate |
| Material | Polypropylene |
| No. of Wells | 96 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?