missing translation for 'onlineSavingsMsg'
Learn More

MAT2A inhibitor 2, MedChemExpress

Product Code. 16494648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494648 10 mg 10mg
16467367 5 mg 5mg
16465486 100 mg 100mg
16420947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494648 Supplier Medchem Express Supplier No. HY112569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MAT2A inhibitor 2 is a methionine adenosyltransferase 2A (MAT2A) inhibitor.

MAT2A inhibitor 2 is a methionine adenosyltransferase 2A (MAT2A) inhibitor.

Specifications

Chemical Name or Material MAT2A inhibitor 2
CAS 13299-99-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H24ClN3O3
Quantity 10 mg
Solubility Information DMSO : 75 mg/mL (205.00 mM; Need ultrasonic)
SMILES O=C1C(C)=CC(C2=CC=CC(OC)=C2)=NN1CCN3CCOCC3.[H]Cl
Molecular Weight (g/mol) 365.85
Formula Weight 365.85
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.