missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MATN4 Monoclonal Antibody (401127)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561402
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Matrilin (MATNs) are a family of non-collagenous extracellular matrix proteins consisting of four known members that have been proposed to play key roles in the formation of both collagen-dependent and collagen-independent filamentous networks. Members of the matrilin family all share a structure made up of von Willebrand factor A domains, epidermal growth factor-like domains and a coiled coil alpha-helical module. MATN1 and MATN3 are expressed mainly in cartilage, while MATN2 and MATN4 occur in a wide variety of extracellular matrices such as embryonic kidney, lung, bone, cartilage and nervous system. The matrilin genes are strictly and differently regulated and their expression may serve as markers for cellular differentiation. MATN4 could serve as an odontoblast differentiation marker, e.g. in odontoblast stem cell research.
Specifications
| MATN4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O89029, O95460 | |
| MATN4 | |
| Mouse myeloma cell line NS0-derived recombinant human Matrilin-4, Gln20-Asp499 (predicted). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 401127 | |
| Unconjugated | |
| MATN4 | |
| HE6WCR54; MAT-4; MATN4; matrilin 4; matrilin-4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 17183, 8785 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction