missing translation for 'onlineSavingsMsg'
Learn More

Matrix Blank: 70% Isooctane/30% Toluene, VHG™

Product Code. 17930606
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17930606 100 mL 100mL
17950606 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17930606 Supplier VHG Labs Supplier No. VHGSISO70TOL30BLK100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Matrix Blank: 70% Isooctane/30% Toluene
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blank
Analyte or Component Names Blank
Shipping Condition Room Temperature
IUPAC Name Blank
Solvent or Matrix Isooctane/Toluene
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.