missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MATRIX Orbital Delta F1.5 Thermoshaker
GREENER_CHOICE

Product Code. 17247509 Shop All IKA Products
Click to view available options
Plug Type:
UK Plug
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17247509

Brand: IKA™ 0020025659

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The MATRIX Orbital Delta F1.5 Thermoshaker mixes and heats with a fixed insert which holds 24 tubes (1.5 mL). Whether its blood samples, pharmaceutical agents, DNA/RNA samples, or ELISA assays, samples with even the smallest volumes mix reliably and completely in all laboratory applications.

  • Large and clear display with a convenient menu structure allows for intuitive operation
  • Speeds of up to 1500 rpm and shaker diameter of 3 mm ensure the liquid mixes quickly and effectively
  • Two different speed modes for gentle heating of the samples
  • Programmable ramps allow for automatic execution of mixing and heating tasks
  • Aluminum die cast housing ensures secure positioning, excellent stability, and general robustness
TRUSTED_SUSTAINABILITY

Specifications

Orbit 3 mm
Load Bearing Capacity (Metric) 0.3 kg
Speed Control Accuracy ±30 rpm
Speed Range 0 to 1500 rpm
Voltage 100/240 V
Temperature Range (Metric) 100°C (Max.)
Wattage 240 W
Product Type Orbital Shaker with Heating
Certifications/Compliance CE
Plug Type UK Plug
Temperature Accuracy ±0.5°C
Dimensions (L x W x H) 320 x 200 x 120 mm
Weight (Metric) 9.2 kg
Controller Type Touch Key Interface
Model MATRIX Orbital Delta F1.5
Display TFT LCD
Humidity Range 80%
Platform Style Fixed Insert for 24 1.5 mL Tubes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.