missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mavorixafor trihydrochloride, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Description
Mavorixafor trihydrochloride (AMD-070 trihydrochloride) is a potent, selective and orally available CXCR4 antagonist, with an IC50 value of 13 nM against CXCR4 125I-SDF binding, and also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with an IC50 of 1 and 9 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Mavorixafor trihydrochloride |
| Physical Form | Solid |
| CAS | 2309699-17-8 |
| Color | Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H30Cl3N5 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | AMD-070 trihydrochloride |
| Solubility Information | DMSO : 150 mg/mL (326.90 mM; Need ultrasonic) ∣H2O : 100 mg/mL (217.93 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction