missing translation for 'onlineSavingsMsg'
Learn More

Mavorixafor trihydrochloride, MedChemExpress

Product Code. 16477836
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477836

Brand: Medchem Express HY50101A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mavorixafor trihydrochloride (AMD-070 trihydrochloride) is a potent, selective and orally available CXCR4 antagonist, with an IC50 value of 13 nM against CXCR4 125I-SDF binding, and also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with an IC50 of 1 and 9 nM, respectively.

Mavorixafor trihydrochloride (AMD-070 trihydrochloride) is a potent, selective and orally available CXCR4 antagonist, with an IC50 value of 13 nM against CXCR4 125I-SDF binding, and also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with an IC50 of 1 and 9 nM, respectively.

Specifications

Chemical Name or Material Mavorixafor trihydrochloride
Physical Form Solid
CAS 2309699-17-8
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H30Cl3N5
For Use With (Application) COVID-19-anti-virus
Synonym AMD-070 trihydrochloride
Solubility Information DMSO : 150 mg/mL (326.90 mM; Need ultrasonic) ∣H2O : 100 mg/mL (217.93 mM; Need ultrasonic)
SMILES [H]Cl.[H]Cl.[H]Cl.NCCCCN(CC1=NC2=CC=CC=C2N1)[C@@H]3C4=NC=CC=C4CCC3
Molecular Weight (g/mol) 458.86
Formula Weight 458.86
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.