missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBD1 Recombinant Rabbit Monoclonal Antibody (24GB405)

Rabbit Recombinant Monoclonal Antibody
198.00 € - 564.00 €
Specifications
| Antigen | MBD1 |
|---|---|
| Clone | 24GB405 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30274805
![]() |
Invitrogen™
MA557470 |
100 μL |
564.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275771
|
Invitrogen™
MA557471 |
20 μL |
198.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MBD1 Recombinant Monoclonal Antibody for Western Blot, Flow
Methyl-CpG-binding domain protein 1 (MBD1) is a member of a family of mammalian proteins that specifically bind methylated regions of DNA. It is a transcriptional repressor that binds CpG islands in promoters where the DNA is methylated at position 5 of cytosine within CpG dinucleotides. Binding is abolished by the presence of 7-mG that is produced by DNA damage by methylmethanesulfonate (MMS). MBD1 acts as a transcriptional repressor and plays a role in gene silencing by recruiting AFT7IP, which in turn recruits factors such as the histone methyltransferase SETDB1. Isoform 1 and isoform 2 can also repress transcription from unmethylated promoters.Specifications
| MBD1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| MBD1 | |
| CXXC3; CXXC-type zinc finger protein 3; Mbd1; methyl-CpG binding domain protein 1; methyl-CpG binding domain protein 1 isoform PCM1; methyl-CpG-binding domain protein 1; Methyl-CpG-binding protein MBD1; PCM1; Protein containing methyl-CpG-binding domain 1; RFT; the regulator of fibroblast growth factor 2 (FGF-2) transcription | |
| MBD1 | |
| Primary | |
| Affinity chromatography |
| 24GB405 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UIS9 | |
| 291439, 4152 | |
| A synthesized peptide derived from human MBD1 (aa 550-605). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title