missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBL1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148564
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Regulator of genomic integrity and cellular response to DNA damage acting through ATR-mediated checkpoint signaling. Necessary for centrosome duplication. It functions as a molecular scaffold facilitating the interaction of PLK4 and CENPJ, two molecules involved in centriole formation.
Specifications
| MBL1 | |
| Polyclonal | |
| Unconjugated | |
| MBD3 | |
| AI181826; AU019209; CpG binding domain 3; Mbd3; methyl-CpG binding domain protein 3; methyl-CpG-binding domain protein 3; Methyl-CpG-binding protein MBD3 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 17192 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Z2D8 | |
| MBD3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MBL-1 (R&D Systems, Catalog # 2077-MB), Ser18-Ala239. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction