Learn More
MBOAT7 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26646-1-AP-20UL
This item is not returnable.
View return policy
Description
The MBOAT7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MBOAT7. This antibody recognizes human, mouse antigen. The MBOAT7 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MBOAT7 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| BB1, h mboa 7, hMBOA 7, LENG4, LPIAT, LPLAT 7, LRC4, Lyso PI acyltransferase, Lysophospholipid acyltransferase 7, MBOA7, MBOAT7, OACT7 | |
| 20 μL | |
| 79143 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |