missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (7D2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-05204-0.025ML
This item is not returnable.
View return policy
Description
MBP Monoclonal specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| MBP | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:5000-1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Mouse | |
| 18.5/21.5 kDa | |
| 0.025 mL | |
| Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
| 4155 | |
| Rat, Human, Mouse, Equine, Pig, Bovine | |
| Ascites |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry | |
| 7D2 | |
| Unconjugated | |
| 50% PBS, 50% Glycerol with 5mM Sodium Azide | |
| MBP | |
| Purified myelin basic protein isolated from bovine brain, epitope maps to the peptide AEGQRPGFGYGGRASDYKSAHKGFKGVDAQGTLSKIFKLG, amino acids 145-184 of the human 21.5kDa sequence. | |
| Affinity purified | |
| RUO | |
| Primary | |
| The MBP Antibody (7D2) antibody binds only the 21.5kDa and 18.5kDa rat MBP isotypes, but all four isotypes of human and bovine MBP. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction