missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MC3R Polyclonal Antibody

Product Code. 16586481
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16586481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16586481 Supplier Invitrogen™ Supplier No. PA588171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MNASCCLPSV QPTLPNGSEH LQAPFFSNQS SSAFCEQVFI KPEVFLSLGI VSLLENILVI LAVVRNGNLH SPMYFFLCSL AVADMLVSVS NALETIMIAI VHSDYLTFED QFIQHMDNIF; Positive Samples: Mouse brain, Mouse liver, Mouse kidney, Rat brain; Cellular Location: Cell membrane, Multi-pass membrane protein.

This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MC3R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MC3R
Gene Accession No. P32244, P33033
Gene Alias BMIQ9; MC3; MC3 receptor; MC3R; MC3-R; melanocortin 3 receptor; melanocortin receptor 3; OB20; obesity quantitative trait locus; OQTL
Gene Symbols MC3R
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17201, 29310
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.