missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM/CD146 Antibody (P1H12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
264.00 € - 441.00 €
Specifications
| Antigen | MCAM/CD146 |
|---|---|
| Clone | P1H12 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry < = 0.25 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18292473
|
Novus Biologicals
NBP1-43346-0.025MG |
0.025 mg |
264.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18716483
|
Novus Biologicals
NBP1-43346-0.1mg |
0.1 mg |
441.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human, Mouse, Canine, Rabbit, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| MCAM/CD146 | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Canine, Rabbit, Rat (Negative) | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
| MCAM | |
| IgG1 κ | |
| Affinity Purified | |
| Does not cross react with Rat. |
| P1H12 | |
| Western Blot 1:100-1:2000, Flow Cytometry < = 0.25 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 4162 | |
| The immunogen for this antibody was CD146. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title