missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MCDB 153 Medium (Modified)

Product Code. 31908457 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908457 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908457 Supplier Sartorius Supplier No. 010591A
 Restricted Item
Estimated Shipment: 19-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

It is a modification of Ham's nutrient mixture F-12 designed for the growth of non-transformed cell types in a serum-free formulation.

  • The medium contains supplements that optimize the growth of specific cell types.
  • MCDB 153 medium is suitable for human epidermal keratinocyte culture, clonal growth of chicken embryo fibroblasts, and Chinese Hamster Ovary (CHO) cells using low levels of Fetal Bovine Serum Protein (FBSP), trace elements, or no serum protein.
  • Modified as follows:
    • With L-Glutamine
    • With Phenol Red
    • With Sodium Bicarbonate
    • With Sodium Pyruvate
    • With Hydrocortisone, T3, Testosterone, Insulin, Transferrin, and Sodium Selenite
    • With MEM-NEAA
    • With Glucose

Specifications

Form Liquid
Product Type Ham's F-12 Cell Culture Media
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.