Learn More
Invitrogen™ MCHR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532917
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gibbon, Monkey, Bovine, Dog, Panda (100%); Mouse, Rat, Horse, Pig (94%); Hamster, Elephant (89%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.
Specifications
| MCHR1 | |
| Polyclonal | |
| Unconjugated | |
| Mchr1 | |
| AW049955; G protein coupled receptor 24; G protein-coupled receptor 24; GPCR24; GPR24; Gpr24-9 protein coupled receptor; G-protein coupled receptor 24; MCH 1 receptor; MCH receptor 1; MCH1; Mch1r; MCH-1R; MCHR; MCHR1; MCH-R1; MCHR-1; melanin concentrating hormone receptor 1; melanin-concentrating hormone receptor 1; SLC1; SLC-1; Somatostatin receptor-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2847, 403563, 540119 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q99705 | |
| Mchr1 | |
| Synthetic 18 amino acid peptide from C-terminus of human MCHR1. | |
| 50 μg | |
| Primary | |
| Human, Canine, Monkey, Bovine | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.