missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCM5 Recombinant Rabbit Monoclonal Antibody (24GB4965)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557467
This item is not returnable.
View return policy
Description
MCM5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
Specifications
| MCM5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MCM5 | |
| AA617332; AI324988; AL033333; Cdc46; CDC46 homolog; DNA replication licensing factor MCM5; MCM5; MCM5 minichromosome maintenance deficient 5, cell division cycle 46; Mcmd5; MGC5315; minichromosome maintenance complex component 5; minichromosome maintenance deficient 5 (cell division cycle 46); minichromosome maintenance deficient 5, cell division cycle 46; minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae); P1-CDC46 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17218, 291885, 4174 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4965 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P33992, P49718 | |
| MCM5 | |
| A synthesized peptide derived from human MCM5 (aa 695-708). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction