Learn More
Invitrogen™ MCM5 Recombinant Rabbit Monoclonal Antibody (24GB4965)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557466
 View more versions of this product
Product Code. 30275028
  • 564.00 € / 100µL
Description
MCM5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MCM5
Recombinant Monoclonal
Unconjugated
MCM5
AA617332; AI324988; AL033333; Cdc46; CDC46 homolog; DNA replication licensing factor MCM5; MCM5; MCM5 minichromosome maintenance deficient 5, cell division cycle 46; Mcmd5; MGC5315; minichromosome maintenance complex component 5; minichromosome maintenance deficient 5 (cell division cycle 46); minichromosome maintenance deficient 5, cell division cycle 46; minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae); P1-CDC46
Rabbit
Affinity Chromatography
RUO
17218, 291885, 4174
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4965
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P33992, P49718
MCM5
A synthesized peptide derived from human MCM5 (aa 695-708).
100 μL
Primary
Human, Mouse, Rat