missing translation for 'onlineSavingsMsg'
Learn More

MCPA Methyl Ester, TRC

Product Code. 30477660
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477660 1 g 1g
30467434 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477660 Supplier Toronto Research Chemicals Supplier No. TRCM1997151G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2436-73-9
Molecular Formula C10 H11 Cl O3
Molecular Weight (g/mol) 214.65
Synonym Acetic acid, 2-(4-chloro-2-methylphenoxy)-, methyl ester, Acetic acid, (4-chloro-2-methylphenoxy)-, methyl ester (9CI), Acetic acid, [(4-chloro-o-tolyl)oxy]-, methyl ester (6CI,7CI,8CI), (4-Chloro-2-methylphenoxy)acetic acid methyl ester, 2-Methyl-4-chlorophenoxyacetic acid methyl ester, MCPA methyl ester, Methyl 4-chloro-2-methylphenoxyacetate
IUPAC Name methyl 2-(4-chloro-2-methylphenoxy)acetate
SMILES COC(=O)COc1ccc(Cl)cc1C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H11ClO3/c1-7-5-8(11)3-4-9(7)14-6-10(12)13-2/h3-5H,6H2,1-2H3
Formula Weight 214.0397
Chemical Name or Material MCPA-methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.