Learn More
MCT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20139-1-AP-20UL
 View more versions of this product
Product Code. 16899634
  • 145.00 € / 20µL
Description
SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
Specifications
Specifications
MCT1
Polyclonal
Unconjugated
SLC16A1
MCT, MCT 1, MCT1, Monocarboxylate transporter 1, SLC16A1
Rabbit
Antigen Affinity Chromatography
RUO
6566
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P53985
SLC16A1
MCT1 Fusion Protein Ag14098
20 μL