missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MDH2 Recombinant Rabbit Monoclonal Antibody (K01_1T73)

Product Code. p-200114081
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824661 100 μL
30830450 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824661 Supplier Invitrogen™ Supplier No. MA563502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MDH2 Recombinant Monoclonal Antibody for IHC (P), Western Blot

Malate dehydrogenase catalyzes the reversible oxidation of malate to oxaloacetate, utilizing the NAD/NADH cofactor system in the citric acid cycle. The protein encoded by this gene is localized to the mitochondria and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDH2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1T73
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene MDH2
Gene Accession No. P04636, P08249, P40926
Gene Alias malate dehydrogenase 2; malate dehydrogenase 2, NAD (mitochondrial); malate dehydrogenase precursor (EC 1.1.1.37); malate dehydrogenase, mitochondrial; MDH; Mdh2; Mdh-2; MGC:3559; mitochondrial malate dehydrogenase 2, NAD; MMDH; M-MDH; Mor1; Mor-1; testicular tissue protein Li 120
Gene Symbols MDH2
Host Species Rabbit
Immunogen A synthetic peptide of human MDH2.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17448, 4191, 81829
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.