missing translation for 'onlineSavingsMsg'
Learn More

Mdivi-1, MedChemExpress

Product Code. p-200096500
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499427 5 mg 5mg
16452957 50 mg 50mg
16446976 100 mg 100mg
16446898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499427 Supplier Medchem Express Supplier No. HY15886

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mdivi-1 is a selective dynamin-related protein 1 (Drp1) inhibitor. Mdivi-1 is a mitochondrial division/mitophagy inhibitor.

Mdivi-1 is a selective dynamin-related protein 1 (Drp1) inhibitor. Mdivi-1 is a mitochondrial division/mitophagy inhibitor.

Specifications

Chemical Name or Material Mdivi-1
CAS 338967-87-6
Color Light Brown
Purity Grade Notes Research
Molecular Formula C15H10Cl2N2O2S
Quantity 5 mg
Synonym Mitochondrial division inhibitor 1
Solubility Information DMSO : 160 mg/mL (452.98 mM; Need ultrasonic)
SMILES O=C1N(C2=C(Cl)C=C(Cl)C(OC)=C2)C(S)=NC3=C1C=CC=C3
Molecular Weight (g/mol) 353.22
Formula Weight 353.22
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.