missing translation for 'onlineSavingsMsg'
Learn More

MDK83190, MedChemExpress

Product Code. 16497548
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497548 10 mg 10mg
16427496 100 mg 100mg
16413577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497548 Supplier Medchem Express Supplier No. HY18633
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MDK83190 is a potent apoptosis activator, induces Apaf-1 oligomerization, increases procaspase-9 processing and subsequent caspase-3 activation in a cyto c-dependent Manner.

MDK83190 is a potent apoptosis activator, induces Apaf-1 oligomerization, increases procaspase-9 processing and subsequent caspase-3 activation in a cyto c-dependent Manner.

Specifications

Chemical Name or Material MDK83190
Physical Form Solid
CAS 79183-19-0
Color Orange
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H9Cl2NO2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 46 mg/mL (150.26 mM)
SMILES O=C1N(CC2=CC=C(Cl)C(Cl)=C2)C3=C(C=CC=C3)C1=O
Molecular Weight (g/mol) 306.14
Formula Weight 306.14
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.