missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDR1/ABCB1 Antibody (6G11F8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00 € - 552.00 €
Specifications
| Antigen | MDR1/ABCB1 |
|---|---|
| Clone | 6G11F8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18161118
|
Novus Biologicals
NBP2-37290 |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683026
|
Novus Biologicals
NBP2-37290-0.025ml |
0.025 mL |
203.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| MDR1/ABCB1 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human | |
| ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
| ABCB1 | |
| IgG1 | |
| Affinity Purified |
| 6G11F8 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Cancer, Membrane Trafficking and Chaperones | |
| PBS with 0.05% Sodium Azide | |
| 5243 | |
| Purified recombinant fragment of human ABCB1 (AA: 632-693) expressed in E. Coli. | |
| Primary | |
| 141.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title