missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ MEBM™ Mammary Epithelial Cell Growth Basal Medium, Phenol Red Free

Product Code. 11695430
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11695430 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11695430 Supplier Lonza™ Supplier No. CC3153
  • ONLINE EXCLUSIVE PRICE
    562.00 €
    468.55 € / Pack of 1
    Save 93.45 €
    16% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Basal medium that can be used with 1 kit of CC-4136 (mammary epithelial cell growth medium SingleQuots™ Supplements and Growth Factors).

  • Basal medium.
  • Phenol Red free
  • Can be used with 1 kit of CC-4136 (mammary epithelial cell growth medium SingleQuots™ Supplements and Growth Factors.
  • Optimized for the growth of mammary epithelial cells in a serum-free environment.
  • This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Product Type Basal Medium
Without Additives No Phenol Red
Quantity 500 mL
pH 7.25 to 7.65
Color Clear
Content And Storage Store in dark at 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.