missing translation for 'onlineSavingsMsg'
Learn More

Meclofenoxate hydrochloride, MedChemExpress

Product Code. 16457366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16457366 Supplier Medchem Express Supplier No. HY17555
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Meclofenoxate hydrochloride, an ester of dimethylethanolamine (DMAE) and 4-chlorophenoxyacetic acid (pCPA), has been shown to improve memory, have a mentally stimulating effect, and improve general cognition.

Meclofenoxate hydrochloride, an ester of dimethylethanolamine (DMAE) and 4-chlorophenoxyacetic acid (pCPA), has been shown to improve memory, have a mentally stimulating effect, and improve general cognition.

Specifications

Chemical Name or Material Meclofenoxate hydrochloride
CAS 3685-84-5
Color White
Purity Grade Notes Research
Molecular Formula C12H17Cl2NO3
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (169.97 mM) ∣H2O : 33.33 mg/mL (113.30 mM; Need ultrasonic)
SMILES ClC1=CC=C(OCC(OCCN(C)C)=O)C=C1.Cl
Molecular Weight (g/mol) 294.17
Formula Weight 294.17
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.