missing translation for 'onlineSavingsMsg'
Learn More

Mecoprop, TRC

Product Code. 30472497
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472497 2.5 g 2.50g
30466049 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30472497 Supplier Toronto Research Chemicals Supplier No. TRCM2030502.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-65-2
Molecular Formula C10 H11 Cl O3
Molecular Weight (g/mol) 214.6455
Synonym Propionic acid, 2-[(4-chloro-o-tolyl)oxy]- (6CI,8CI), (±)-2-(4-Chloro-2-methylphenoxy)propanoic acid, (±)-2-(4-Chloro-2-methylphenoxy)propionic acid, (±)-Mecoprop, 2-(2-Methyl-4-chlorophenoxy)propanoic acid, 2-(2-Methyl-4-chlorophenoxy)propionic acid, 2-(4-Chloro-2-methylphenoxy)propanoic acid, 2-(4-Chloro-2-methylphenoxy)propionic acid, 2-(4-Chloro-o-tolyloxy)propionic acid, 2-(p-Chloro-o-tolyloxy)propionic acid, 2M4KhP, Anicon B, Anicon P, CMPP, Celatox CMPP, Compitox, Isocarnox, MCPP, MCPP (herbicide), Mechlorprop, Mecoprop, Morogal, NSC 60282, Okultin MP, Rankotex, SYS 67 Mecmin, U 46 KV Fluid, α-(2-Methyl-4-chlorophenoxy)propionic acid
IUPAC Name 2-(4-chloro-2-methylphenoxy)propanoic acid
SMILES CC(Oc1ccc(Cl)cc1C)C(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C10H11ClO3/c1-6-5-8(11)3-4-9(6)14-7(2)10(12)13/h3-5,7H,1-2H3,(H,12,13)
Formula Weight 214.0397
Chemical Name or Material Mecoprop
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.