Learn More
Invitrogen™ MED13L Recombinant Rabbit Monoclonal Antibody (HL2911)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556786
Product Code. 30275035
  • 500.00 € / 100µL
Description
MED13L Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene is a subunit of the Mediator complex, a large complex of proteins that functions as a transcriptional coactivator for most RNA polymerase II-transcribed genes. The encoded protein is involved in early development of the heart and brain. Defects in this gene are a cause of transposition of the great arteries, dextro-looped (DTGA).[provided by RefSeq, Jul 2010].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MED13L
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Q71F56
MED13L
Synthetic peptide of human MED13L.
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL2911
Unconjugated
MED13L
2210413I17Rik; 6330591G05Rik; 9030618F05Rik; Kiaa1025; Med13l; mediator complex subunit 13 like; mediator complex subunit 13-like; Mediator of RNA polymerase II transcription subunit 13-like; mKIAA1025; MRFACD; PROSIT240; Thrap2; thyroid hormone receptor associated protein 2; thyroid hormone receptor-associated protein 2; Thyroid hormone receptor-associated protein complex 240 kDa component-like; TRAP240L
Rabbit
Protein A
RUO
23389
-20°C, Avoid Freeze/Thaw Cycles