missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99047-100ul
This item is not returnable.
View return policy
Description
MED4 Polyclonal antibody specifically detects MED4 in Human samples. It is validated for Western Blot
Specifications
| MED4 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| Activator-recruited cofactor 36 kDa component, DRIP36mediator of RNA polymerase II transcription, subunit 4 homolog (S. cerevisiae), FLJ10956, HSPC126vitamin D receptor interacting protein, mediator complex subunit 4TRAP/SMCC/PC2 subunit p36 subunit, subunit 4 homolog, TRAP36 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MED4. | |
| 100 μg | |
| Signal Transduction | |
| 29079 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction