missing translation for 'onlineSavingsMsg'
Learn More

Medetomidine hydrochloride, MedChemExpress

Product Code. 16487278
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487278 10 mg 10mg
16453297 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487278 Supplier Medchem Express Supplier No. HY17034B10mg
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Medetomidine hydrochloride is an agonist of adrenergic alpha-2 receptor, which is used in veterinary medicine for its analgesic properties.

Medetomidine hydrochloride is an agonist of adrenergic alpha-2 receptor, which is used in veterinary medicine for its analgesic properties.

Specifications

Chemical Name or Material Medetomidine hydrochloride
Physical Form Solid
CAS 86347-15-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H17ClN2
For Use With (Application) Neuroscience-Neuromodulation
Synonym MPV785
Solubility Information DMSO : 100 mg/mL (422.40 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (422.40 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (211.20 mM)
SMILES CC(C1=CN=CN1)C2=CC=CC(C)=C2C.Cl
Molecular Weight (g/mol) 236.74
Formula Weight 236.74
Percent Purity 99.92%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.