missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Medium 199 with Earle's Salt (E199), with L-glutamine, without Sodium Bicarbonate

Product Code. 11400275 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400275 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11400275 Supplier MP Biomedicals™ Supplier No. 091020122
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medium 199 with Earle's Salt (E199), with L-glutamine, without Sodium Bicarbonate is a completely defined nutritional source for cell culture.

  • Medium 199, originally described by Morgan and his colleagues (1950), is a completely defined nutritional source for cell culture.
  • Their investigations demonstrated that cell growth could be measured in this medium.
  • It has broad species applicability including the culturing of non-transformed cell types.
  • It may be used for vaccine production and the in vitro cultivation of rat lens tissues and primary mouse pancreatic epithelial explants.
  • It is available in either Earle's or Hanks' salts.

Specifications

Form Powder
Quantity 10 L

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.