missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 199, Hanks' Balanced Salts, powder

Product Code. 11513387 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 L
1 L
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11513387 10 L 10 L
10165142 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11513387 Supplier Gibco™ Supplier No. 10012037
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medium 199, Hanks' Balanced Salts, powder

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. This Medium 199 formulation contains Hanks' salts for use without CO2.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing
Gibco™ Medium 199 is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Medium 199 contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This Medium 199 formulation does not require a CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Medium 199 Hanks' Balanced Salts
With Additives Glutamine, Hanks’ Balanced Salt solution (HBSS), HEPES, Phenol Red
Without Additives No Sodium Bicarbonate
For Use With (Application) For cell culture, mammalian cell culture, vaccine production and In-vitro cultivation
Quantity 10 L
Certifications/Compliance ISO 13485 standards, FDA
pH 7 to 7.4
Content And Storage 2 to 8°C
Description Hanks' Balanced Salts, Medium 199, 10L, Powdered Form, L-Glutamine, HEPES Buffer, 2 to 8°C Storage Temperature, 36 Months Shelf Life, For Mammalian Cell Culture, Vaccine Production and In-vitro Cultivation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.