missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 199, HEPES

Product Code. 11580596 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11580596 10 x 500 mL Pack of 10
11570596 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11580596 Supplier Gibco™ Supplier No. 22340087
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medium 199, HEPES

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ Medium 199 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Medium 199 product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Medium 199 contains no proteins, lipids, or growth factors. Medium 199 uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type Medium 199
With Additives Glutamine, HEPES, Phenol Red, Sodium Pyruvate
For Use With (Application) Mammalian cell culture
Quantity 10 x 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7.1 to 7.3
Regulatory Status In-vitro Diagnostic Use
Content And Storage 2 to 8°C
Description Medium 199, 500mL, 1 X Concentration, Liquid Form, With L-glutamine, Phenol Red, HEPES, Sodium Pyruvate, 7.1 to 7.3 pH, 12 Months Shelf Life, 270 to 330mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.